Learn More
C5b-9 Antibody (aE11), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP1-05120F
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
C5b-9 Monoclonal antibody specifically detects C5b-9 in Human, Porcine, Bacteria, Equine, Baboon samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Western Blot (Negative)
Specifica
| C5b-9 | |
| Monoclonal | |
| FITC | |
| MAC, Membrane attack complex, sC5-b9, sC5b-9, TCC, Terminal complement complex | |
| Purified C5b-9 | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| aE11 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human, Pig, Bacteria, Equine, Baboon | |
| Purified |