missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcineurin B Antibody (BE2.1), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-08451APC
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Calcineurin B Monoclonal antibody specifically detects Calcineurin B in Human, Xenopus samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)
Specifica
| Calcineurin B | |
| Monoclonal | |
| APC | |
| calcineurin subunit B type 1, CNA2, CNB1, Protein phosphatase 2B regulatory subunit 1, protein phosphatase 2B regulatory subunit B alpha, protein phosphatase 3 (formerly 2B), regulatory subunit B (19kD), alpha isoform(calcineurin B, type I), protein phosphatase 3 (formerly 2B), regulatory subunit B, 19kDa, alpha isoform(calcineurin B, type I), protein phosphatase 3 (formerly 2B), regulatory subunit B, alpha isoform, Protein phosphatase 3 regulatory subunit B alpha isoform 1, protein phosphatase 3, regulatory subunit B, alpha, type I (19kDa) | |
| Recombinant full-length human Calcineurin B protein (Uniprot: P63098) | |
| 0.1 mL | |
| Protein Phosphatase, Signal Transduction, Wnt Signaling Pathway | |
| 5534 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| BE2.1 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Xenopus species | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto