Learn More
Fistreem International™ Calypso Ultrapure Water Still
Click to view available options
Portata uscita:
2 L/hr.
4 L/hr.
Tensione:
220 V
240 V
Wattage:
1.8 kW
3.0 kW
Dimensione della confezione:
Pezzo
Descrizione
Calypso water stills combine many of the proven features of the higher specification Cyclon still range with a budget design concept. Available with outputs of 2L/hr and 4L/hr, both units produce the highest quality pyrogen free water achieved by a unique cyclonic effect vapour steam trap. A transparent acrylic cover protects the glassware and allows inspection. Heating is provided by silica sheathed elements. The stills are supplied ready for connection to a mains tap supply but, where preferred, can be connected to a pre-treated water feed (optional feed solenoid valve required). In the pre-treated mode, purified water is fed directly to the boiler and tap water is used for cooling purposes only. The still is supplied with a polyethylene reservoir level sensor, providing automatic shut-off for both power and input water whenever the reservoir level is full. The still restarts automatically whenever the reservoir level drops. Overnight running can be carried out safely and reliably, providing a reserve of freshly produced water at the beginning of the day. For easy cleaning, the still is fitted with a boiler drain valve. Cleaning solution is introduced into the boiler via the constant level device. No dismantling is necessary. A high temperature cut-off protects the heaters in the event of water failure. An automatic flow controller governs the rate of water consumption and allows the unit to be used on a wide tap water pressure range. The stills may be bench or wall mounted and are supplied with all necessary connection hoses and wall brackets. Calypso stills are offered with an optional matching reservoir tank. Automatic controls allow up to 30L distilled water to be maintained. The reservoir has a fast flow tap, air filter and sight tube and may be bench or wall mounted.
Specifica
Specifica
| Portata uscita | 2 L/hr. |
| Dimensioni (L x P x A) | 790 x 260 x 325 mm |
| Tensione | 240 V |
| Wattage | 1.8 kW |
| Inlet Pressure | 70 to 560 kPa 10 to 80 lbf.in-² |