Learn More
Eurofins DiscoverX CaM Kinase IIβ Protein, Active
Description
CAMK2B (Calcium/Calmodulin Dependent Protein Kinase II Beta) belongs to the serine/threonine protein kinase family and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. Calcium signaling is crucial for several aspects of plasticity at glutamatergic synapses. In mammalian cells, the enzyme is composed of four different chains: alpha, beta, gamma, and delta. The product of this gene is a beta chain. It is possible that distinct isoforms of this chain have different cellular localizations and interact differently with calmodulin.
GenBank AF081572
Specifications
Specifications
| Formulazione | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Peso molecolare | 39.6 kDa |
| Quantità | 10 μg |
| Specie | Human |
| Recombinant | Recombinant |
| Tag proteine | N-terminal 6His |
| Sistema di espressione | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥65% |
| Proteine | CaM Kinase IIβ , active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.