Learn More
Invitrogen™ CaMKI Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA1543
Descrizione
PA1-543 detects calciμm/calmodulin-dependent kinase I (CaMKI) from rat and mouse brain extract. PA1-543 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~43 kDa protein representing CaMKI from rat and mouse brain extract. PA1-543 immunizing peptide corresponds to amino acid residues 1-18 from human CaMKI protein. PA1-543 immunizing peptide (Cat. # PEP-134) is available for use in neutralization and control experiments.
CAMK1 (CaMK1) is a serine/threonine kinase that is a member of the calcium/calmodulin-dependent protein kinase family. It is involved in cell differentiation, nervous system development, and signal transduction. CAMK1 is expressed in many tissues and is a component of a calmodulin-dependent protein kinase cascade. Calcium/calmodulin directly activates calcium/calmodulin-dependent protein kinase I by binding to the enzyme and indirectly promotes the phosphorylation and synergistic activation of the enzyme by calcium/calmodulin-dependent protein kinase I kinase.
Specifica
| CaMKI | |
| Polyclonal | |
| Unconjugated | |
| CAMK1 | |
| AI505105; calcium/calmodulin dependent protein kinase I; calcium/calmodulin-dependent protein kinase I; calcium/calmodulin-dependent protein kinase type 1; CaM kinase I; CaM kinase I alpha; Camk1; CAMK1-PEN; Camki; caM-KI; CaMKIalpha; CaMKI-alpha; D6Ertd263e; G alpha interacting protein; Gaip; MGC120317; MGC120318; regulator of G-protein signalling 19 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 171503, 52163, 8536 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| Q14012, Q63450, Q91YS8 | |
| CAMK1 | |
| Synthetic Peptide: A(1) V E G P R W K Q A E D I R D I Y D(18). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.