missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaMKII beta Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11533-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The Ca2+/calmodulin-dependent protein kinases (CaM kinases) comprise a structurally related subfamily of serine/threonine kinases which include CaMKI, CaMKII and CaMKIV. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+ and calmodulin (CaM) and has been implicated in regulation of the cell cycle and transcription. There are four CaMKII isozymes, designated alpha, beta, gamma and delta, which may or may not be co-expressed in the same tissue type. CaMKII alpha is autophosphorylated on Thr 286 upon the binding of the Ca2+/CaM complex to the autoinhibitory domain of CaMKII. This process is called Ca2+/CaM trapping, which is thought to be involved in the synaptic encoding of information.Specifica
| CaMKII beta | |
| Polyclonal | |
| Unconjugated | |
| CAMK2B | |
| CaM kinase II subunit beta, CAM2, CaMK II subunit beta, CAMK2, CAMK2B, CAMKB, CaMKII beta | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12323, 24245, 816 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P28652, Q13554 | |
| CAMK2B | |
| CaMKII beta Fusion Protein Ag2119 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto