Learn More
Paragon Scientific™ Cannon-Fenske Routine Viscometer for Transparent Liquids
Click to view available options
Intervallo viscosità:
0.5 to 2 cSt
0.8 to 4 cSt
1.6 to 8 cSt
10,000 to 50,000 cSt
100 to 500 cSt
1600 to 8000 cSt
20 to 100 cSt
20,000 to 100,000 cSt
240 to 1200 cSt
3 to 15 cSt
4000 to 20,000 cSt
50 to 250 cSt
500 to 2500 cSt
7 to 35 cSt
Dimensione della confezione:
Pezzo
Descrizione
- High Quality glass capillary viscometer
- Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifica
Specifica
| Intervallo viscosità | 20 to 100 cSt |
| Calibrated | No |
| Costante approssimata | 0.1 |
| Numeri ASTM | D 445, D 446 |
| Certificazione/Conformità | ASTM D 445, D 446 and ISO 3104, 3105 |
| Size | 200 |
4 week lead-time