Learn More
Thermo Scientific™ Thermo Scientific™ Dual Pocket Specimen Transport Bags
Click to view available options
Product Type:
Specimen Transport Bag
Dimensione della confezione:
pezzi 1000
Descrizione
Features:
- Bags with dual-pocket front pocket for specimens and back pocket for requisition or sensitive documents
- Removable liner exposes robust adhesive for sealing both compartments of the bag
- Printed biohazard symbol and instructions for use
Ordering Information:
Customized printed biohazard bags available, minimum quantity required.
Includes:
Transport bag available with or without an absorbent pad.
Specifica
Specifica
| Product Type | Specimen Transport Bag |
| Descrizione | Dual Pocket Specimen Transport Bag with Absorbent Sheet |
| Altezza (sistema imperiale) | 9 in. |
| Include | A front pocket for specimens and a back pocket for documentation, an integrated adhesive seal, and the bag is printed with a Biohazard symbol and instructions for use |
| N. per custodia | 1000/Cs. |
| Larghezza (sistema imperiale) | 6 in. |