Learn More
Thermo Scientific™ Thermo Scientific™ Specimen Collection Beakers
Click to view available options
Capacità (sistema metrico):
103.5 mL
236 mL
Capacità (sistema imperiale):
3.5 oz.
8 oz.
Include:
Pour Spout
Temperature Strip
Dimensione della confezione:
pezzi 350
pezzi 500
Descrizione
Thermo Scientific™ Collection Beakers are offered in two volumes, 3 oz. and 8 oz., to support a variety of sample collection needs. The beakers feature pour spouts for easy sample transfer.
Features:
- Pour spout for easy transfer of sample into smaller containers
- With or without temperature indicator strips
Specifica
Specifica
| Product Type | Specimen Collection Cup |
| Materiale | Polypropylene |
| Capacità (sistema imperiale) | 3.5 oz. |
| Capacità (sistema metrico) | 103.5 mL |
| Descrizione | Collection Cup with pour spout; 3.5 oz. (103.5 mL) |
| Include | Pour Spout |
| N. per custodia | 350/Cs. |