Learn More
Erlab™ Captair™ Filtration Column for Fume Hoods
Click to view available options
Product Type:
Horizontal Filtration Column
Vertical Filtration Column
Tipo di filtro:
Securifilter AS Filter
Securifilter BE Filter
Include:
01 Sampling Port
02 Sampling Port, Fan Failure Alarm
03 Sampling Port, Fan Failure Alarm
04 Sampling Port
Dimensione della confezione:
Pezzo
Descrizione
- Modular filtration column
- Handlings in an ultra-clean environment
- Serial prefilter (optimizes the HEPA H14 filter lifetime)
- Particulate HEPA H14 filtration
- Optional carbon filtration = clean the air polluted by ambient air VOCs
- Equipped with a HEPA H14 filter, guarantees 99.995% filtration efficiency for particles larger than 0.1μm (according to the MPPS method set forth in the EN 1822-1 standard)
- Ultra-clean air entering the enclosure meets ISO class 5* (EN ISO 14-644 standard) requirements, which corresponds to American class 100 (i.e., less than 100 particles per cubic foot > 0.5 μm) and to class A and B of the GMP guide published by the European Union for the pharmaceutical industry
- Items located into the enclosure are therefore protected from any external contaminants
- HEPA filtered enclosure
Applications:
- Non-pathogenic cell cultures
- In-vitro cultures
- Microbiology (Non-pathogenic)
- Homeopathic preparations in pharmacies
- Electronics
- Optics, etc.
Specifica
Specifica
| Tipo di filtro | Securifilter AS Filter |
| Altezza (sistema metrico) | 587 mm |
| Include | 02 Sampling Port, Fan Failure Alarm |
| Profondità (sistema metrico) | 537 mm |
| Product Type | Vertical Filtration Column |
| Larghezza (sistema metrico) parte esterna | 200 mm |
| Da utilizzare con (applicazione) | For storage with organic vapors mainly for under bench safety cabinets |