Learn More
Erlab™ Captair Smart Glass Work Surface with Retention Tray
Codice prodotto. 15575164
Click to view available options
Da utilizzare con (apparecchiatura):
Smart 321 Fume Hood
Smart 391 Fume Hood
Smart 392 Fume Hood
Smart 481 Fume Hood
Smart 483 Fume Hood
Smart 633 Fume Hood
Smart 714 Fume Hood
Dimensione della confezione:
Pezzo
Codice prodotto. 15575164
Marca: Erlab™ I0166020001
Erlab™ Captair Smart Work Surface with Retention Tray is designed to be used with the range of Captair Smart Ductless Filtering Fume Hoods.
  • Tempered enameled glass worktop
  • Includes retention tray filtering fume hoods
  • Jointed frame
  • Ergonomic arm rest for a comfortable working position
Specifica
Da utilizzare con (apparecchiatura) Smart 714 Fume Hood
Include Retention Tray
Profondità (sistema metrico) 585 mm
Descrizione Jointed Frame
Larghezza (sistema metrico) 1703 mm
Product Type Work Surface
Materiale Tempered Glass