Learn More
Thermo Scientific™ CaptureSelect™ Biotin Anti-EPO Conjugate
Sfoglia Tutto Thermo Scientific ProdottiDescrizione
CaptureSelect Biotin Anti-EPO Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for both recombinant and natural-sourced human erythropoietin (EPO). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.
Applications for CaptureSelect Biotin Anti-EPO Conjugate include ELISA, western blot, Gyrolab™-based immunoassays, and use of label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Specifica
Specifica
| Content And Storage | Store at -5°C to -30°C. |
| Label Type | Biotin and Other Haptens |
| Linea di prodotti | CaptureSelect |
| Product Type | Ligand Conjugates |
| Target Species | Human |
| Tipo di legante | Antibody |
| Quantità | 500 μg |
| Shipping Condition | Approved for shipment on Wet or Dry Ice |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.