Learn More
Carbic anhydride, 99+%, Thermo Scientific Chemicals
28.00€ - 176.00€
Identificatori chimici
CAS | 129-64-6 |
---|---|
Formula molecolare | C9H8O3 |
Molecular Weight (g/mol) | 164.16 |
Numero MDL | MFCD00151106,MFCD00069013,MFCD00630580,MFCD00151106 |
InChI Key | KNDQHSIWLOJIGP-UHFFFAOYNA-N |
Sinonimo | cis-5-norbornene-endo-2,3-dicarboxylic anhydride |
PubChem CID | 126963424 |
IUPAC Name | 4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione |
SMILES | O=C1OC(=O)C2C3CC(C=C3)C12 |
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificatori chimici
129-64-6 | |
164.16 | |
KNDQHSIWLOJIGP-UHFFFAOYNA-N | |
126963424 | |
O=C1OC(=O)C2C3CC(C=C3)C12 |
C9H8O3 | |
MFCD00151106,MFCD00069013,MFCD00630580,MFCD00151106 | |
cis-5-norbornene-endo-2,3-dicarboxylic anhydride | |
4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione |
Specifica
Carbic anhydride | |
162.0°C to 165.0°C | |
1.0800g/mL | |
Balance: himic acid | |
1.4365 | |
15, 1797 | |
cis-5-norbornene-endo-2,3-dicarboxylic anhydride | |
KNDQHSIWLOJIGP-UHFFFAOYNA-N | |
4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione | |
126963424 | |
99+% |
129-64-6 | |
White | |
Authentic | |
C9H8O3 | |
MFCD00151106,MFCD00069013,MFCD00630580,MFCD00151106 | |
1.08 | |
Solubility in water: <0,01g/100g. Other solubilities: soluble in acetone,benzene,ethyl ether,ethanol,decomposes with alkalies | |
O=C1OC(=O)C2C3CC(C=C3)C12 | |
164.16 | |
164.16 | |
Crystalline Powder |
Sicurezza e movimentazione
GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Causes serious eye damage.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of s
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 204-957-7
RUO – Research Use Only