Learn More
Swann-Morton™ Carbon Steel Individual Peel Pack, Non-Sterile Scalpel Blades, No. 21
Click to view available options
Quantità:
100/Box
Dimensione della confezione:
pezzi 100
Descrizione
Large version of No. 10 blade with a curved, cutting edge and a flat, unsharpened back edge.
- Large version of No. 10 blade.
- Curved cutting edge.
- Flat, unsharpened back edge.
- Larger than the No. 20.
- Smaller than the No. 22.
- Fits handles 4, 4L, 4 graduated, and 6B.
Specifica
Specifica
| Materiale della lama | Carbon Steel |
| Quantità | 100/Box |
| Tipo di imballaggio | Individually Packaged |