missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase IX/CA9 Antibody (4F12), Novus Biologicals™
Rabbit Monoclonal Antibody
412.00€ - 640.00€
Specifica
| Antigene | Carbonic Anhydrase IX/CA9 |
|---|---|
| Clone | 4F12 |
| Diluizione | ELISA, Immunohistochemistry 1:50-1:200 |
| Applicazioni | ELISA, Immunohistochemistry |
| Classificazione | Monoclonal |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
30025668
|
Novus Biologicals
NBP3-26468-50ul |
50 μL |
412.00€
50µL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
30039895
|
Novus Biologicals
NBP3-26468-100ul |
100 μL |
640.00€
100µL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
Carbonic Anhydrase IX/CA9 Monoclonal antibody specifically detects Carbonic Anhydrase IX/CA9 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifica
| Carbonic Anhydrase IX/CA9 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CAIX, CA-IX, Carbonate dehydratase IX, carbonic anhydrase 9, carbonic anhydrase IX, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MN, P54/58N, PMW1, RCC-associated antigen G250, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250 | |
| A synthesized peptide derived from Human Carbonic Anhydrase IX/CA9 [UniProt Q16790] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 4F12 | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, HIF Target Genes, Hypoxia, Lipid and Metabolism, Metastasis, Renal Cell Carcinoma Cell Markers | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 768 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title