missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 10/p23/17/p12 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14311-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. This protein cleaves and activates caspases 3 and 7, and the protein itself is processed by caspase 8. Mutations in this gene are associated with apoptosis defects seen in type II autoimmune lymphoproliferative syndrome. Three alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifica
| Caspase 10/p23/17/p12 | |
| Polyclonal | |
| Unconjugated | |
| CASP10 | |
| ALPS2, Apoptotic protease Mch 4, CASP 10, CASP10, Caspase 10, Caspase 10/p23/17/p12, Caspase10, FLICE2, ICE like apoptotic protease 4, MCH4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 843 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q92851 | |
| CASP10 | |
| Caspase 10/p23/17/p12 Fusion Protein Ag5511 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto