missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Caspase 7 p11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578924
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: HELA whole cell, MCF-7 whole cell, Rat Liver Tissue, Rat Kidney Tissue. IHC: Human Intestinal Cancer Tissue.
This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. The precursor of this caspase is cleaved by caspase 3 and 10. It is activated upon cell death stimuli and induces apoptosis. Alternative splicing results in four transcript variants, encoding three distinct isoforms.
Specifications
| Caspase 7 p11 | |
| Polyclonal | |
| Unconjugated | |
| CASP7 | |
| AI314680; Apoptotic protease Mch-3; Casp7; CASP-7; caspase 7; caspase 7, apoptosis-related cysteine peptidase; caspase 7, apoptosis-related cysteine protease; caspase-7; Caspase-7 subunit p11; Caspase-7 subunit p20; CMH-1; cysteine protease LICE2; ICE-IAP3; ICE-LAP3; ICE-like apoptotic protease 3; LICE2; mCASP-7; MCH3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12369, 64026, 840 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| P55210, P97864 | |
| CASP7 | |
| A synthetic peptide corresponding to a sequence in the middle region of human Caspase-7 (P11) (237-255aa RGSWFVQALCSILEEHGKD). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction