Learn More
Epredia™ Cassette II Slotted Tissue Cassettes
Sfoglia Tutto Epredia Prodotti
Click to view available options
Colore:
Beige
Blue
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Dimensione della confezione:
pezzi 1000
Descrizione
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifica
Specifica
| Stile pori | Slots |
| Chiusura inclusa | Yes |
| Quantità | 1000/Cs. |
| Tipo di chiusura | Separate Lid, Front Hinge |
| Colore | Orange |
| Product Type | Slotted Cassette |