Learn More
CCS Viscosity Standard, ASTM D 5293 and SAE Specification J317, Paragon Scientific™
Click to view available options
Quantità:
500 mL
Dimensione della confezione:
500mL
Descrizione
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Full Traceability to National Standards
- Ensures full compliance to ASTM & IP test method protocol
- Manufactured in the United Kingdom
- 2 year shelf life
Specifica
Specifica
| Imballaggio | Glass Amber Bottle |
| Certificato di analisi | Yes |
| Forma fisica | Liquid |
| Quantità | 500 mL |
Nominal values quoted. Certified Values will vary from batch to batch.