Learn More
Invitrogen™ CD14 Monoclonal Antibody (MEM-18), APC

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA110095
Descrizione
This antibody reacts with CD14, a 53-55 kDa GPI (glycosylphosphatidylinositol)-linked extracellular membrane glycoprotein expressed on monocytes, macrophages and weakly on granulocytes; also expressed by most tissue macrophages. In human, the epitope recognized by MEM-18 is located between amino acids 57-64.
CD14 is a 55 kDa GPI-anchored glycoprotein that is constitutively expressed on the surface of mature monocytes, macrophages, and neutrophils. CD14 also serves as a multifunctional lipopolysaccharide receptor, and is released to the serum both as a secreted and enzymatically cleaved GPI-anchored form. CD14 binds lipopolysaccharide molecule in a reaction catalyzed by lipopolysaccharide-binding protein (LBP), an acute phase serum protein. The soluble sCD14 can discriminate slight structural differences between lipopolysaccharides and is important for neutralization of serum allochthonous lipopolysaccharides by reconstituted lipoprotein particles. Further, CD14 has been shown to bind apoptotic cells, and can affect allergic, inflammatory and infectious processes. Alternative splicing results in multiple transcript variants encoding the same CD14 isoform. Diseases associated with CD14 dysfunction include mycobacterium chelonae infection and Croup.
Specifica
| CD14 | |
| Monoclonal | |
| 0.045 mg/mL | |
| PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
| P08571 | |
| Cd14 | |
| A crude mixture of human urinary proteins precipitated by ammonium sulphate from the urine of a patient suffering from proteinuria. | |
| 100 Tests | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| MEM-18 | |
| APC | |
| Cd14 | |
| CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
| Mouse | |
| Size-exclusion chromatography | |
| RUO | |
| 929 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.