missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2 Antibody (RPA-2.10), Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-25200
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CD2 Monoclonal specifically detects CD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Block/Neutralize.
Specifica
| CD2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 0.1-0.25 ug per 1x10^6 cells. Use reported by customer review, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Block/Neutralize | |
| CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This product reacts with Human. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Blocking Assay | |
| RPA-2.10 | |
| Unconjugated | |
| NP_001758 | |
| CD2 | |
| A recombinant protein was used as the immunogen for the CD2 antibody. | |
| 0.1 mg | |
| Adaptive Immunity, Apoptosis, Immunology | |
| 914 | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto