missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Mouse anti-Baboon, Human, Rhesus, , Brilliant Ultraviolet 737, Clone: 2H7, BD Horizon™
Mouse Monoclonal Antibody
Marca: BD Biosciences 612849
Dettagli aggiuntivi : Peso : 0.00020kg
Descrizione
CD20 Monoclonal antibody specifically detects CD20 in Baboon,Human,Rhesus, samples. It is validated for Flow Cytometry.Specifica
CD20 | |
Monoclonal | |
Brilliant Ultraviolet 737 | |
Mouse | |
Affinity Purified | |
RUO | |
Baboon, Human, Rhesus Monkey | |
IgG2b κ |
Flow Cytometry | |
2H7 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human 6.16c1.3 B cell line | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |