Learn More
CD24 Rat anti-Mouse, APC, Clone: M1/69, Proteintech
Rat Monoclonal Antibody
Marca: Proteintech APC-65121-100UG
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The CD24 antibody from Proteintech is a rat monoclonal antibody generated with n/a of mouse CD24. This antibody recognizes mouse antigen. The CD24 antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifica
| CD24 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Rat | |
| Primary | |
| Mouse |
| Flow Cytometry | |
| M1/69 | |
| APC | |
| 100 μg | |
| 12484 | |
| IgG2b |