missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Mouse anti-Human, BB515, Clone: CD28.2, BD Horizon™
Mouse Monoclonal Antibody
Marca: BD Biosciences 564492
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CD28 Monoclonal antibody specifically detects CD28 in Human samples. It is validated for Flow cytometry.
Specifica
| CD28 | |
| Monoclonal | |
| Brilliant Blue 515 (BB515) | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 100 Tests | |
| Cell Biology | |
| 940 | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. | |
| IgG1 κ |
| Flow Cytometry | |
| CD28.2 | |
| Vol. Per test: 5μL | |
| CD28 antigen; T44; Tp44; TP44 | |
| Human CD28 Transfected Cell Line | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Reseach Use Only.
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto