missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD299 (DC-SIGN/L) Polyclonal Antibody

CD299 (DC-SIGN/L) Polyclonal Antibody
Marca: Thermo Scientific PA560068
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CD299 (DC-SIGN/L) Polyclonal Antibody for IHC
Specifica
| CD299 (DC-SIGN/L) | |
| Polyclonal | |
| Unconjugated | |
| CLEC4M | |
| CD209 antigen-like protein 1; Cd209b; CD209b antigen; CD209L; CD209L1; CD299; CD299 antigen; CLEC4M; C-type lectin domain family 4 member M; C-type lectin domain family 4, member M; DC-SIGN2; DCSIGNR; DC-SIGNR; DC-SIGN-related protein; Dendritic cell-specific ICAM-3-grabbing non-integrin 2; HP10347; liver/lymph node-specific ICAM-3 grabbing non-integrin; liver/lymph node-specific ICAM-3-grabbing non-integrin; LSIGN; L-SIGN; mannose binding C-type lectin DC-SIGNR; MGC129964; MGC47866 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10332 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H2X3 | |
| CLEC4M | |
| Recombinant protein corresponding to Human CLEC4M. Recombinant protein control fragment (Product #RP-98388). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Ser du en mulighed for forbedring?Del en indholdskorrektion