missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse anti-Human , APC-R700, Clone: OKT3, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566797
Dettagli aggiuntivi : Peso : 0.00020kg
Descrizione
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow CytometrySpecifica
CD3 | |
Monoclonal | |
APC-R700 | |
CD3E; CD3e; T-cell surface antigen T3/Leu-4 epsilon; T3E; TCRE | |
Sheep Erythrocyte Rosette-purified Human T Cells | |
RUO | |
Human | |
IgG2a κ |
Flow Cytometry | |
OKT3 | |
Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |