missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse anti-Human , Clone: OKT3, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566685
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for T-Cell Activation, Flow Cytometry
Specifica
| CD3 | |
| Monoclonal | |
| No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
| Mouse | |
| 1 mg | |
| Primary | |
| Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. This preparation contains no preservatives, thus it should be handled under aseptic conditions. |
| T-Cell Activation, Flow Cytometry | |
| OKT3 | |
| CD3E; CD3e; T-cell surface antigen T3/Leu-4 epsilon; T3E; TCRE | |
| Sheep Erythrocyte Rosette-purified Human T Cells | |
| RUO | |
| Human | |
| IgG2a κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto