Learn More
CD3 Mouse anti-Human, CoraLite 488, Clone: OKT3, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL488-65133-100TESTS
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The CD3 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD3. This antibody recognizes human antigen. The CD3 antibody has been validated for the following applications: FC analysis. This antibody is coralite™488.
Specifica
| CD3 | |
| Monoclonal | |
| 5 μL/Test | |
| Mouse | |
| Primary | |
| Human |
| Flow Cytometry | |
| OKT3 | |
| CoraLite 488 | |
| 100 Tests | |
| 916 | |
| IgG2a |