missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD36 Antibody (D-2712), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NB110-59724V
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CD36 Monoclonal specifically detects CD36 in Mouse, Rat, Human (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifica
| CD36/SR-B3 | |
| Monoclonal | |
| DyLight 405 | |
| CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor | |
| Mouse | |
| 75 kDa | |
| 0.1 mL | |
| Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Stem Cell Markers | |
| 948 | |
| Mouse, Rat, Human (Negative) | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| D-2712 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CD36 | |
| Adenovirus expressing recombinant mouse CD36. [Uniprot: Q08857] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse and rat. Does not react with human. | |
| Store at 4C in the dark. | |
| IgA κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto