Learn More
CD4 Rat anti-Mouse, PE, Clone: GK1.5, Proteintech
Rat Monoclonal Antibody
Marca: Proteintech PE-65104-100UG
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The CD4 antibody from Proteintech is a rat monoclonal antibody generated with mouse ctl clone v4 of mouse CD4. This antibody recognizes mouse antigen. The CD4 antibody has been validated for the following applications: FC analysis. This antibody is pe.
Specifica
| CD4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Rat | |
| Primary | |
| Mouse |
| Flow Cytometry | |
| GK1.5 | |
| PE | |
| 100 μg | |
| 12504 | |
| IgG2b |