missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 v4 Mouse anti-Human, BV750, Clone: L120, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 747216
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CD4 v4 Monoclonal antibody specifically detects CD4 v4 in Human samples. It is validated for Flow Cytometry
Specifica
| CD4 v4 | |
| Monoclonal | |
| Brilliant Violet 750 | |
| L3T4; T-cell surface antigen T4/Leu-3; W3/25; CD4 antigen (p55) | |
| Human Peripheral Blood Lymphocytes | |
| RUO | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| L120 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV750 under optimal conditions that minimize unconjugated dye and antibody. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto