missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD41/CD61 Monoclonal Antibody (CO.35E4)
Mouse Monoclonal Antibody
Marca: Invitrogen™ MA528370
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Mouse anti Sheep CD41/CD61 antibody, clone CO.35E4 recognizes ruminant CD41/CD61, also known as GpIIb/IIIa. CD41/CD61 is a calcium-complex dependent heterodimer and is expressed almost exclusively on platelets and on their precursors, the megakaryocytes.
CD41 is a transmembrane glycoprotein (integrin family) composed of two chains GPIIb alpha (heavy chain; 120 kDa) and GPIIb beta (light chain; 23 kDa). CD61 non-covalently associates with CD41.
Specifica
| CD41/CD61 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| 0 | |
| ITGA2B, ITGB3 | |
| Sheep leucocytes. | |
| 100 μg | |
| Primary | |
| Canine, Rabbit, Bovine, Sheep, Feline, Horse, Goat | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunoprecipitation | |
| CO.35E4 | |
| Unconjugated | |
| ITGA2B | |
| alphaIIb protein; antigen CD61; BDPLT16; BDPLT2; CD41; CD41B; CD61; glycoprotein GPIIb; glycoprotein GPIIIa; glycoprotein IIb; glycoprotein IIIa; GP2B; GP3A; GPalpha IIb; GPIIb; GPIIIa; GT; GTA; HPA3; INTB3; integrin; integrin alpha 2b; integrin alpha 2b precursor; Integrin alpha-2b; integrin alpha-IIb; Integrin alpha-IIb heavy chain; Integrin alpha-IIb light chain, form 1; Integrin alpha-IIb light chain, form 2; Integrin beta; integrin beta 3; integrin beta 3 subunit; integrin beta3; integrin beta-3; integrin beta-3 subunit; integrin subunit alpha 2b; integrin subunit alpha IIb; integrin subunit beta 3; integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41); integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41B); integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); ITGA2B; ITGA2B-like protein; ITGAB; ITGB3; platelet fibrinogen receptor, alpha subunit; platelet glycoprotein IIb; platelet glycoprotein IIb of IIb/IIIa complex; platelet glycoprotein IIIa; platelet glycoprotein IIIa integrin subunit beta 3; platelet membrane glycoprotein IIb; Platelet membrane glycoprotein IIIa; platelet-specific antigen BAK; PPP1R93; protein phosphatase 1, regulatory subunit 93 | |
| Mouse | |
| Protein A | |
| RUO | |
| 100008734, 100008735, 100009697, 100009709, 100423047, 101092159, 101100930, 101115719, 102169417, 102183451, 282642, 403788, 403789, 515011 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto