Learn More
CD43 Mouse anti-Human, CoraLite 488, Clone: 2A11D6, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL488-66224-100UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The CD43 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD43. This antibody recognizes human antigen. The CD43 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifica
| CD43 | |
| Monoclonal | |
| 2000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Immunofluorescence | |
| 2A11D6 | |
| CoraLite 488 | |
| 100 μL | |
| 6693 | |
| IgG2b |