Learn More
CD45.1 Mouse anti-Mouse, CoraLite 647, Clone: A20, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL647-65092-100TESTS
Descrizione
The CD45.1 antibody from Proteintech is a mouse monoclonal antibody generated with sjl mouse thymocytes and splenocytes of mouse CD45.1. This antibody recognizes mouse antigen. The CD45.1 antibody has been validated for the following applications: FC analysis. This antibody is coralite™647.
Specifica
| CD45.1 | |
| Monoclonal | |
| 5 μL/Test | |
| Mouse | |
| Primary | |
| Mouse |
| Flow Cytometry | |
| A20 | |
| CoraLite 647 | |
| 100 Tests | |
| 19264 | |
| IgG2a |