missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45R (B220) Rat anti-Mouse, Human, BUV737, Clone: RA3-6B2, BD Horizon™
Rat Monoclonal Antibody
Marca: BD Biosciences 612838
Dettagli aggiuntivi : Peso : 0.00030kg
Descrizione
CD45R (B220) Monoclonal antibody specifically detects CD45R (B220) in Mouse,Human samples. It is validated for Flow Cytometry.Specifica
CD45R (B220) | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
CD45R (B220) | |
Refrigerate (2 to 8°C) |
Flow Cytometry | |
RA3-6B2 | |
Brilliant Ultraviolet 737 | |
50 μg | |
Primary | |
Mouse, Human | |
IgG2a κ |