Learn More
CD45RO Mouse anti-Human, BB515, Clone: UCHL1, BD Horizon™
Mouse Monoclonal Antibody
195.81€ - 709.41€
Specifica
Antigene CD45RO
Clone UCHL1
Applicazioni Flow Cytometry
Classificazione Monoclonal
Coniugato Brilliant Blue 515 (BB515)
Visualizza altre specifiche
Prodotti 2
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
15803709
Vedere i documenti
BD Biosciences
564530
25 Tests
ONLINE EXCLUSIVE PRICE
196.00€
195.81€ / Pezzo
Risparmia 0.19€
0.10% Sconto
Visualizzare prodotti alternativi
Risparmia fino a 
»
15893699
Vedere i documenti
BD Biosciences
564529
100 Tests
ONLINE EXCLUSIVE PRICE
710.00€
709.41€ / Pezzo
Risparmia 0.59€
0.08% Sconto
Visualizzare prodotti alternativi
Risparmia fino a 
»
Descrizione
Descrizione
CD45RO Monoclonal antibody specifically detects CD45RO in Human samples. It is validated for Flow cytometry.
CD45RO Monoclonal antibody specifically detects CD45RO in Human samples. It is validated for Flow cytometry.
Specifica
Specifica
CD45RO
Flow Cytometry
Brilliant Blue 515 (BB515)
Mouse
Cell Biology
Aqueous buffered solution containing ≤0.09% sodium azide.
Human IL-2-dependent T-cell line
Primary
Vol. Per test: 5μL
UCHL1
Monoclonal
Purified
RUO
Human
CD45R; PTPRC; LCA; Leukocyte common antigen; GP180; LY5; T200
IgG2a κ
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Documenti
Product Certifications
For Research Use Only.