Learn More
CD52 Mouse anti-Human, Unconjugated, Clone: 1B10E8, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66784-1-IG-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The CD52 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD52. This antibody recognizes human antigen. The CD52 antibody has been validated for the following applications: WB, ELISA analysis.
Specifica
| CD52 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 1B10E8 | |
| Unconjugated | |
| 20 μL | |
| 1043 | |
| IgG1 |