Learn More
Novus Biologicals™ CD59 Overexpression Lysate
Codice prodotto. 18083858 Sfoglia Tutto Bio Techne Prodotti
Click to view available options
Quantità:
0.1 mg
Dimensione della confezione:
0.10mg
Codice prodotto. 18083858
Marca: Novus Biologicals™ NBP210121
The CD59 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Specifica
Linea cellulare The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_97675. The protein contains a C-terminal DDK tag.
Da utilizzare con (applicazione) Western Blot
Quantità 0.1 mg
Tampone di conservazione RIPA buffer
Purezza Protein
Test di specificità Homo sapiens CD59, mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Sinonimo 16.3A5, 1F5, 1F5 antigen, 20 kDa homologous restriction factor, CD59 antigen, CD59 antigen, complement regulatory protein, CD59 glycoprotein, CD59 molecule, complement regulatory protein, EJ16, EJ30, EJ30, EL32 and G344), EL32, FLJ38134, FLJ92039, G344, HRF20, HRF-20, human leukocyte antigen MIC11, Ly-6-like protein, lymphocytic antigen CD59/MEM43, MACIF, MAC-inhibitory protein, MAC-IP, MEM43, MEM43 antigen, membrane attack complex (MAC) inhibition factor, Membrane attack complex inhibition factor, Membrane inhibitor of reactive lysis, MGC2354, MIC11MSK21, MIN1, MIN2, MIN3, MIRL, p18-20, protectin, surface anitgen recognized by monoclonal 16.3A5, T cell-activating protein
Protein State Native
For Research Use Only