Learn More
CD73 Mouse anti-Human, PE, Clone: AD2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech PE-65162-100TESTS
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The CD73 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD73. This antibody recognizes human antigen. The CD73 antibody has been validated for the following applications: FC analysis. This antibody is pe.
Specifica
| CD73 | |
| Monoclonal | |
| 10 μL/Test | |
| Mouse | |
| Primary | |
| Human |
| Flow Cytometry | |
| AD2 | |
| PE | |
| 100 Tests | |
| 4907 | |
| IgG1 κ |