missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD81 Monoclonal Antibody (Eat2), PE

Armenian Hamster Monoclonal Antibody
224.00€ - 564.00€
Specifica
| Antigene | CD81 |
|---|---|
| Clone | Eat2 |
| Concentrazione | 0.1 mg/mL |
| Content And Storage | 4°C, do not freeze |
| Applicazioni | Flow Cytometry |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
15394211
![]() |
Invitrogen™
MA517941 |
50 μg |
224.00€
50µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
15304221
|
Invitrogen™
MA517942 |
300 μg |
564.00€
300µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
CD81 Monoclonal Antibody for Flow
CD81 (TAPA-1, target of anti-proliferative antibody-1) is a member of the tetraspanin family, is expressed on virtually all nucleated cells, but above all on germinal center B cells. CD81 forms complexes with other tetraspanin proteins, integrins, coreceptors, MHC class I and II molecules, and influences adhesion, morphology, activation, proliferation and differentiation of B, T cells. In muscles, CD81 promotes cell fusion and myotube maintenance. CD81 has been also identified as a receptor for the hepatitis C virus. Like members of the tetraspanin family that include CD9, CD37, CD53, CD63, and CD82, CD81 is a cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. CD81 is a cell surface glycoprotein that is known to complex with integrins. CD81 appears to promote muscle cell fusion and support myotube maintenance. CD81 associates with CD19, CD21, Leu 13, and integrins on cell membrane and acts as a receptor for the envelope protein E2 of chronic hepatitis C virus. Antibodies to CD81 have anti-proliferative effects on different lymphoid cell lines, particularly those derived from large cell lymphomas. CD81 is also localized in the tumor-suppressor gene region and is a candidate gene for malignancies.Specifica
| CD81 | |
| 0.1 mg/mL | |
| Flow Cytometry | |
| PE | |
| Armenian Hamster | |
| RUO | |
| PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
| P35762, Q62745 | |
| 12520, 25621 | |
| Mouse CD81 (TAPA-1). | |
| Antibody |
| Eat2 | |
| 4°C, do not freeze | |
| Monoclonal | |
| Liquid | |
| IgG | |
| Mouse, Rat | |
| CD81 | |
| 26 kDa cell surface protein TAPA-1; CD 81 antigen; Cd81; CD81 antigen; CD81 antigen (target of antiproliferative antibody 1); CD81 molecule; CVID6; S5.7; Tapa1; Tapa-1; Target of the antiproliferative antibody 1; Tetraspanin28; tetraspanin-28; Tspan28; Tspan-28 | |
| CD81 | |
| Primary | |
| Protein G |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto