Learn More
CD86 Mouse anti-Human, PE, Clone: 2331 (FUN-1), BD
Mouse Monoclonal Antibody
180.00€ - 393.82€
Specifica
| Antigene | CD86 |
|---|---|
| Clone | 2331 (FUN-1) |
| Applicazioni | Flow Cytometry |
| Classificazione | Monoclonal |
| Coniugato | PE |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
15800639
|
BD Biosciences
560957 |
25 Tests |
180.00€
Pezzo |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
15828338
|
BD Biosciences
555658 |
100 Tests |
495.00€ 393.82€ / Pezzo Risparmia 101.18€ 20% Sconto |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
The 2331 (FUN-1) monoclonal antibody specifically recognizes a 75 kDa transmembrane cell surface protein, CD86 (B70/B7-2), expressed primarily on monocytes, dendritic cells and activated B cells. Competitive binding assays demonstrate that, while both 2331 (FUN-1) and IT2.2 (anti-CD86, Cat. No. 555663) antibodies specifically recognize the same molecule, they react with different epitopes. CD86 is the second ligand for CD28 and CTLA-4 and may play an important role in co-stimulation of T cells in primary immune response. The 2331 (FUN-1) antibody blocks the costimulatory activity of CD86 when tested in functional studies.
This antibody is routinely tested on Daudi cells by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifica
| CD86 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| B7.2; B7-2; B-lymphocyte activation antigen B7-2; B70; BU63; CD28LG2; LAB72 |
| 2331 (FUN-1) | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human HBL-1 Cell Line | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.