missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90/Thy1 Antibody (7E1B11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Marca: Novus Biologicals NBP2-37330
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CD90/Thy1 Monoclonal specifically detects CD90/Thy1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifica
| CD90/Thy1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen | |
| Mouse | |
| 18 kDa | |
| 0.1 mL | |
| Hematopoietic Stem Cell Markers, Hepatic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
| 7070 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 7E1B11 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| THY1 | |
| Purified recombinant fragment of human THY1 (AA: 17-132) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto