missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD94 Monoclonal Antibody (18d3)

Rat Monoclonal Antibody
Marca: Invitrogen™ MA528670
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CD94 Monoclonal Antibody for Flow
Natural killer (NK) cells, like CD94, are a distinct lineage of lymphocytes that mediate cytotoxic activity and secrete cytokines upon immune stimulation. Several genes of the C-type lectin superfamily, including members of the NKG2 family, are expressed by NK cells and may be involved in the regulation of NK cell function. KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus. Three transcript variants encoding two different isoforms have been found for this gene.
Specifica
| CD94 | |
| Monoclonal | |
| 0.5 mg/mL | |
| BBS with no preservative; pH 8.2 | |
| O54707 | |
| KLRD1 | |
| CHO cells transfected with the B6 allele of CD94. | |
| 1 mL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
| Flow Cytometry | |
| 18d3 | |
| Unconjugated | |
| KLRD1 | |
| Cd94; CD94 antigen; CD94 antigen (located within the rat natural killer gene complex); killer cell lectin like receptor D1; Killer cell lectin-like receptor subfamily D member 1; killer cell lectin-like receptor subfamily D, member 1; killer cell lectin-like receptor, subfamily D, member 1; KLRD1; KP43; natural killer cell receptor; natural killer cells antigen CD94; NK cell receptor | |
| Rat | |
| Purified | |
| RUO | |
| 16643 | |
| 4°C | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto