missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD99 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

462.00€ - 824.00€

Specifica

Antigene CD99
Concentrazione 0.057 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applicazioni Immunohistochemistry (Paraffin), Western Blot
Classificazione Polyclonal
Visualizza altre specifiche

Prodotti 2
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Precio Cantidad y disponibilidad  
11501733
Ver documentos
Invitrogen™
PA137370
1 mL
824.00€
1mL
Ver productos alternativos
Ahorre hasta 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
13456857
GREENER_CHOICE
Ver documentos
Invitrogen™
PA532337
500 μL
462.00€
500µL
Ver productos alternativos
Ahorre hasta 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Descripción

Descripción

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is pancreas or Ewing's sarcoma.

CD99 (E2, MIC2) is a transmembrane glycoprotein that is involved in regulation of T cell addhesive properties and programmed cell death distinct from typical apoptosis course. CD99 roles are specific to certain stages of T cell differentiation such as corticothymocytes. CD99R isoform expression is restricted in the haematopoietic system to T, NK and myeloid cells.
Especificaciones

Especificaciones

CD99
-20°C, Avoid Freeze/Thaw Cycles
Polyclonal
Liquid
IgG
Human
CD99
1110061M03Rik; 12E7; 2410026K10Rik; antigen identified by monoclonal 12E7, Y homolog; antigen identified by monoclonal antibodies 12E7, F21 and O13; Cd99; CD99 antigen; CD99 molecule; cell surface antigen 12E7; cell surface antigen HBA-71; cell surface antigen O13; D4; E2 antigen; HBA71; MIC2; MIC2 (monoclonal antibody 12E7); MIC2X; MIC2Y; MSK5X; paired immunoglobin-like type 2 receptor-ligand; pilr-1; Pilrl; PILR-L; Protein MIC2; surface antigen MIC2; T-cell surface glycoprotein E2
CD99
Primary
Antigen affinity chromatography
0.057 mg/mL
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
RUO
PBS with 1% BSA and 0.08% sodium azide; pH 7.6
P14209
4267
Recombinant full length human CD99 protein.
Antibody
Videos
SDS
Documentos

Documentos

Product Certifications
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.