missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC42 Antibody (M430), Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-23213
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CDC42 Monoclonal antibody specifically detects CDC42 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifica
| CDC42 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Mouse | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| M430 | |
| Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:200 | |
| CDC42Hs, cell division control protein 42 homolog, cell division cycle 42 (GTP binding protein, 25kDa), cell division cycle 42 (GTP-binding protein, 25kD), dJ224A6.1.1 (cell division cycle 42 (GTP-binding protein, 25kD)), dJ224A6.1.2 (cell division cycle 42 (GTP-binding protein, 25kD)), G25K, G25K GTP-binding protein, growth-regulating protein, GTP-binding protein, 25kD, small GTP binding protein CDC42 | |
| Clone (M430) was generated from a rat recombinant Cdc42 protein. This sequence is highly conserved in human and mouse Cdc42. [UniProt# Q8CFN2] | |
| 0.1 mL | |
| Breast Cancer, Cancer, Phospho Specific, Signal Transduction | |
| 998 | |
| Store at -20°C. | |
| IgM |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto