missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDH17 Monoclonal Antibody (4F6H5)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA530203
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CDH17/Cadherin-17.
CDH17 gene is a member of the cadherin superfamily, genes encoding calcium-dependent, membrane-associated glycoproteins. The encoded protein is cadherin-like, consisting of an extracellular region, containing 7 cadherin domains, and a transmembrane region but lacking the conserved cytoplasmic domain. The protein is a component of the gastrointestinal tract and pancreatic ducts, acting as an intestinal proton-dependent peptide transporter in the first step in oral absorption of many medically important peptide-based drugs. The protein may also play a role in the morphological organization of liver and intestine. Alternative splicing results in multiple transcript variants.
Specifica
| CDH17 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q12864 | |
| Cdh17 | |
| Recombinant Human CDH17/Cadherin-17 Protein (Met1-Met787). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| ELISA | |
| 4F6H5 | |
| Unconjugated | |
| Cdh17 | |
| BILL-cadherin; Cadherin; cadherin 17; cadherin 17, LI cadherin (liver-intestine); cadherin-16; cadherin-17; CDH16; CDH17; FLJ26931; HPT1; HPT-1; HPT-1 cadherin; HPT-1/LI; human intestinal peptide-associated transporter HPT-1; human peptide transporter 1; intestinal peptide-associated transporter HPT-1; LI cadherin; LI-cadherin; Liver-intestine cadherin; MGC138218; MGC142024; P130 | |
| Mouse | |
| Protein A | |
| RUO | |
| 1015 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto