Learn More
Eurofins DiscoverX Cdk1/cyclin B Protein, Active
Descrizione
The cyclin-dependent kinases (CDKs), a subfamily of protein kinases, promote cell cycle progression. In mammals, nine different CDKs (cdk 1 to cdk 9; cdk 1 is better known as cdc 2) have been identified. CDKs are regulated on various levels including binding of proteins (cyclins, inhibitors, and assembly factors), protein degradation, transcriptional control, localization, and multiple phosphorylations. This leads to timely regulation of CDK activity, allowing progression from one cell cycle phase to the next. The cdk1 induces entry into M-phase of the cell cycle in eukaryotes.
GenBank NM_001786 / EMBL M25753
Specifica
Specifica
| Formulazione | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Peso molecolare | 35 kDa (CDK1) and 75 kDa (Cyclin B) |
| Quantità | 10 μg |
| Specie | Human |
| Recombinant | Recombinant |
| Tag proteine | C-terminal 6His / N-terminal GST |
| Sistema di espressione | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | Combined ≥63% |
| Proteine | CDK1/Cyclin B, active |
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.