Learn More
Greiner Bio-One CELLCOAT™ 384-well, Poly-L Lysine-Treated, Flat-Bottom Microplate
Click to view available options
Colore:
Black, Clear Bottom
Dimensione della confezione:
pezzi 20
Descrizione
- Improves adhesion of different cell types to polystyrene surfaces
- Reduced-serum or serum-free cultivation
- Increase in isolation and cultivation efficiency
- Ready-to-use products, time-saving
- Consistent quality
Specifica
Specifica
| Formato | Standard |
| Coperchio | With Lid |
| Matrice | 16 x 24 |
| Autoclavabile | Not Autoclavable |
| Imballaggio | 5/Pk., 20/Cs. |
| Binding Type | Non Binding |
| Colore | Black, Clear Bottom |
| Materiale | Polystyrene |
| No. of Wells | 384 |
| Volume (sistema metrico) di lavorazione | 15 to 110 μL |
| Vedi altri risultati |