Learn More
Greiner Bio-One Matracci per colture cellulari Cellcoat™
Click to view available options
Capacità (sistema metrico):
50 mL
250 ml
Include:
Red filter screw cap
Writing area; Red filter screw cap
Dimensione della confezione:
pezzi 5
pezzi 50
Volume (sistema metrico) di lavorazione:
5 to 10 mL
Descrizione
- Promotion of cell adhesion, proliferation and growth of endothelial cell, hepatocytes, muscle cells, pheochromocytoma cells (PC12) and other cell types
- Shelf life: 24 months at room temperature
- Cell cultivation in serum-free or serum-reduced medium
- Rat tail Collagen Type I
- Increase in isolation and cultivation efficiency
- Ready-to-use products: immediate use, time-saving
- Consistent quality
- Storable at room temperature
Specifica
Specifica
| Capacità (sistema metrico) | 250 ml |
| Materiale | Polistirene |
| Include | Writing area; Red filter screw cap |
| Stile collo | Canted |
| N. per custodia | Confezione da 250 |
| N. per confezione | 250 |
| Rivestimento superficiale | Collagen Type I Protein |
| Graduated | Sì |