Learn More
Greiner Bio-One CELLCOAT™ Poly-D-Lysine Protein Coated Flasks
GREENER_CHOICE
Codice prodotto. 11799482
Click to view available options
Area di coltura:
175 cm2
Volume (sistema metrico) di lavorazione:
50 mL
650mL
Dimensione della confezione:
pezzi 40
pezzi 50
Codice prodotto. 11799482
Marca: Greiner Bio-One 661940
Ideal for low-serum, serum-free cultivation and cell differentiation. Greiner Bio-One CELLCOAT™ Poly-D-Lysine Protein Coated Flasks helps in promotion of cell adhesion and proliferation
Poly-D-lysine (PDL) are synthetic molecules which increase cell adhesion to vessels by altering the net charge of the surface without stimulating biological activity.
  • Coated with Poly-D-Lysine synthetic protein (75-150 kDa)
  • For promotion of cell adhesion, proliferation and growth of transfected cell lines, neuronal cell lines, primary neurons and glial cells
  • Ready-to-use products
  • Includes lid with condensation rings
TRUSTED_SUSTAINABILITY
Specifica
Rivestimento superficiale Poly-D-Lysine
Tipo di chiusura Plug Seal Cap
Area di coltura 175 cm2
Volume (sistema metrico) di lavorazione 650mL